1-Methyl-1H-indole-6-carbonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Cyano-1-methylindole
OR8652-1G
APOSOR8652-1GEA
33.5
EUR
APOSOR8652-1G
1-Methyl-1H-indole-6-carbonitrile 97%
1-Methyl-1H-indole-6-carbonitrile
Formel:
C₁₀H₈N₂ Schmelzpunkt: 68…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690254 CAS-Nummer: 20996-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...