1,1,4,4-Tetraphenyl-1,3-butadien 98%

Lieferant: Apollo Scientific
Achtung

OR8656-25G
APOSOR8656-25GEA 306 EUR
APOSOR8656-25G
1,1,4,4-Tetraphenyl-1,3-butadien 98%
1,1,4,4-Tetraphenyl-1,3-butadien

For scintillation work

Formel: (C₆H₅)₂CCHCHC(C₆H₅)₂
Molecular Weight: 358,48 g/mol
Schmelzpunkt: 202…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004766
CAS-Nummer: 1450-63-1
EINECS: 215-914-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR