2,4-Dimethoxy-5-pyrimidinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethoxypyrimidine-5-boronic acid, (2,4-Dimethoxypyrimidin-5-yl)boronic acid, 2,4-Dimethoxy-5-pyrimidylboronic acid

OR8664-1G
APOSOR8664-1GEA 36.4 EUR
APOSOR8664-1G
2,4-Dimethoxy-5-pyrimidinboronsäure
2,4-Dimethoxy-5-pyrimidineboronic acid
Formel: C₆H₉BN₂O₄
Molecular Weight: 183,9577 g/mol
Schmelzpunkt: 113…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01318985
CAS-Nummer: 89641-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR