5-Morpholino-2-thiophenecarbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR8675-1G
OR8675-500MG
APOSOR8675-1GEA
79.9
EUR
APOSOR8675-1G
APOSOR8675-500MG
5-Morpholino-2-thiophenecarbaldehyde 97%
5-Morpholino-2-thiophenecarbaldehyde
Formel:
C₉H₁₁NO₂S Molecular Weight: 197.26 g/mol Schmelzpunkt: 126…128 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02710611 CAS-Nummer: 24372-49-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...