4-(Aminomethyl)-1-methyl-1H-indole 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(1-Methyl-1H-indol-4-yl)methylamine

OR8682-1G OR8682-250MG
APOSOR8682-1GEA 348 EUR
APOSOR8682-1G APOSOR8682-250MG
4-(Aminomethyl)-1-methyl-1H-indole 97%
4-(Aminomethyl)-1-methyl-1H-indole
Formel: C₁₀H₁₂N₂
Molecular Weight: 160.216 g/mol
Schmelzpunkt: 43…47 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08690264
CAS-Nummer: 864264-02-8
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR