(2-Piperidin-1-ylpyridin-4-yl)methylamine

Lieferant: Apollo Scientific
Gefahr

OR8684-1G
APOSOR8684-1GEA 183 EUR
APOSOR8684-1G
(2-Piperidin-1-ylpyridin-4-yl)methylamine
(2-Piperidin-1-ylpyridin-4-yl)methylamine
Formel: C₁₁H₁₇N₃
Molecular Weight: 191,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08690284
CAS-Nummer: 876316-37-9
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR