4-(2-Dimethylaminoethoxy)phenylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-[2-(Dimethylamino)ethoxy]aniline

OR8695-1G
APOSOR8695-1GEA 256 EUR
APOSOR8695-1G
4-(2-Dimethylaminoethoxy)phenylamine 97%
4-(2-Dimethylaminoethoxy)phenylamine
Formel: C₁₀H₁₆N₂O
Molecular Weight: 180,25 g/mol
Schmelzpunkt: 54,5…57.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07801177
CAS-Nummer: 62345-76-0
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR