7-Chloro-4-(piperazin-1-yl)quinoline 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR8697-1G
OR8697-250MG
APOSOR8697-1GEA
0
EUR
APOSOR8697-1G
APOSOR8697-250MG
7-Chloro-4-(piperazin-1-yl)quinoline 97%
7-Chloro-4-(piperazin-1-yl)quinoline
Formel:
C₁₃H₁₄ClN₃ Schmelzpunkt: 113…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01476123 CAS-Nummer: 837-52-5 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...