4'-(Piperidin-4-ylcarbonyl)acetanilide 97%

Lieferant: Apollo Scientific

OR8698-1G OR8698-250MG
APOSOR8698-1GEA 219 EUR
APOSOR8698-1G APOSOR8698-250MG
4'-(Piperidin-4-ylcarbonyl)acetanilide 97%
4'-(Piperidin-4-ylcarbonyl)acetanilide
Formel: C₁₄H₁₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08690322
CAS-Nummer: 124035-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR