(2-Methyl-4-phenyl-1,3-thiazol-5-yl)methanol
Lieferant: Apollo Scientific
OR8704-1G
APOSOR8704-1GEA
232
EUR
APOSOR8704-1G
(2-Methyl-4-phenyl-1,3-thiazol-5-yl)methanol
(2-Methyl-4-phenyl-1,3-thiazol-5-yl)methanol
Formel:
C₁₁H₁₁NOS Schmelzpunkt: 143…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690247 CAS-Nummer: 857284-12-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...