[4-(4-Methylhomopiperazin-1-yl)phenyl]methanol 97%

Lieferant: Apollo Scientific

Synonyme: [4-(4-Methyl-1,4-diazepan-1-yl)phenyl]methanol, [4-(4-Methylperhydro-1,4-diazepin-1-yl)phenyl]methanol

OR8707-1G
APOSOR8707-1GEA 248 EUR
APOSOR8707-1G
[4-(4-Methylhomopiperazin-1-yl)phenyl]methanol 97%
[4-(4-Methylhomopiperazin-1-yl)phenyl]methanol
Formel: C₁₃H₂₀N₂O
Molecular Weight: 220,311 g/mol
Schmelzpunkt: 96,5…97.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08690315
CAS-Nummer: 898289-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR