8-Bromchinolin-2(1H)-on 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8708-1G
OR8708-5G
OR8708-10G
APOSOR8708-1GEA
23.2
EUR
APOSOR8708-1G
APOSOR8708-5G
APOSOR8708-10G
8-Bromchinolin-2(1H)-on 98%
8-Bromchinolin-2(1H)-on
Formel:
C₉H₆BrNO Molecular Weight: 224.0540 g/mol Schmelzpunkt: 185…186 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741409 CAS-Nummer: 67805-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...