8-Bromchinolin-2(1H)-on 98%

Lieferant: Apollo Scientific
Achtung

OR8708-1G OR8708-5G OR8708-10G
APOSOR8708-1GEA 23.2 EUR
APOSOR8708-1G APOSOR8708-5G APOSOR8708-10G
8-Bromchinolin-2(1H)-on 98%
8-Bromchinolin-2(1H)-on
Formel: C₉H₆BrNO
Molecular Weight: 224.0540 g/mol
Schmelzpunkt: 185…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08741409
CAS-Nummer: 67805-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR