(2,4-Diaminopteridin-6-yl)methanol hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: (2,4-Diaminopteridin-6-yl)methanol hydrochloride, 2,4-Diamino-6-(hydroxymethyl)pteridine hydrochloride, 6-(Hydroxymethyl)pteridine-2,4-diamine hydrochloride

OR8715-5G
APOSOR8715-5GEA 108 EUR
APOSOR8715-5G
(2,4-Diaminopteridin-6-yl)methanol hydrochloride
(2,4-Diaminopteridin-6-yl)methanol hydrochloride
Formel: C₇H₉ClN₆O
Molecular Weight: 228,64 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191246
CAS-Nummer: 73978-41-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR