1-Brom-3,5-di-tert-butylbenzol
Lieferant: Apollo Scientific
|
Achtung
|
OR8717-100G
OR8717-25G
OR8717-5G
APOSOR8717-100GEA
163
EUR
APOSOR8717-100G
APOSOR8717-25G
APOSOR8717-5G
1-Brom-3,5-di-tert-butylbenzol
1-Brom-3,5-di-tert-butylbenzol
Formel:
C₁₄H₂₁Br Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00796945 CAS-Nummer: 22385-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...