4-(4-Chlorophenyl)piperidine 95%

Lieferant: Apollo Scientific

OR8735-1G
APOSOR8735-1GEA 115 EUR
APOSOR8735-1G
4-(4-Chlorophenyl)piperidine 95%
4-(4-Chlorophenyl)piperidine
Formel: C₁₁H₁₄ClN
Molecular Weight: 195,69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03077010
CAS-Nummer: 26905-02-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR