(S)-(+)-4-Phenyl-2-oxazolidinon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-4-Phenyloxazolidin-2-one

OR8745-500G
APOSOR8745-500GEA 162 EUR
APOSOR8745-500G
(S)-(+)-4-Phenyl-2-oxazolidinon 98%
(S)-(+)-4-Phenyl-2-oxazolidinon
Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
Schmelzpunkt: 129…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043396
CAS-Nummer: 99395-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR