2-Formylphenylboronsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure

OR8773-100G OR8773-25G OR8773-5G
APOSOR8773-100GEA 143 EUR
APOSOR8773-100G APOSOR8773-25G APOSOR8773-5G
2-Formylphenylboronsäure 97%
2-Formylphenylboronsäure
Formel: C₇H₇BO₃
Molecular Weight: 149.94 g/mol
Schmelzpunkt: 115…120 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00151822
CAS-Nummer: 40138-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR