2-Formylphenylboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Boronobenzaldehyd, (2-Formylphenyl)metaborsäure, Dihydroxy(2-formylphenyl)boran, 2-Formylbenzolboronsäure
OR8773-100G
OR8773-25G
OR8773-5G
APOSOR8773-100GEA
143
EUR
APOSOR8773-100G
APOSOR8773-25G
APOSOR8773-5G
2-Formylphenylboronsäure 97%
2-Formylphenylboronsäure
Formel:
C₇H₇BO₃ Molecular Weight: 149.94 g/mol Schmelzpunkt: 115…120 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00151822 CAS-Nummer: 40138-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...