5-(4-Methylphenyl)isoxazole

Lieferant: Apollo Scientific
Achtung

OR8779-1G
APOSOR8779-1GEA 0 EUR
APOSOR8779-1G
5-(4-Methylphenyl)isoxazole
5-(4-Methylphenyl)isoxazole
Formel: C₁₀H₉NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00030520
CAS-Nummer: 7064-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR