4'-Hydroxy-2'-methoxyacetophenone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Hydroxy-2-methoxyphenyl)ethanone, 4-Acetyl-3-methoxyphenol, 1-(4-Hydroxy-2-methoxyphenyl)ethan-1-one

OR8814-1G OR8814-250MG
APOSOR8814-1GEA 116 EUR
APOSOR8814-1G APOSOR8814-250MG
4'-Hydroxy-2'-methoxyacetophenone
4'-Hydroxy-2'-methoxyacetophenone
Formel: C₉H₁₀O₃
Molecular Weight: 166.174 g/mol
Schmelzpunkt: 136…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038750
CAS-Nummer: 493-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR