4'-Hydroxy-2'-methoxyacetophenone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(4-Hydroxy-2-methoxyphenyl)ethanone, 4-Acetyl-3-methoxyphenol, 1-(4-Hydroxy-2-methoxyphenyl)ethan-1-one
OR8814-1G
OR8814-250MG
APOSOR8814-1GEA
116
EUR
APOSOR8814-1G
APOSOR8814-250MG
4'-Hydroxy-2'-methoxyacetophenone
4'-Hydroxy-2'-methoxyacetophenone
Formel:
C₉H₁₀O₃ Molecular Weight: 166.174 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038750 CAS-Nummer: 493-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...