4-Bromo-3,5-dichloropyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR8846-25G
APOSOR8846-25GEA
74.1
EUR
APOSOR8846-25G
4-Bromo-3,5-dichloropyridine
4-Bromo-3,5-dichloropyridine
Formel:
C₅H₂BrCl₂N Schmelzpunkt: 75…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797978 CAS-Nummer: 343781-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...