4-Methyl-1,3-thiazol-5-carbaldehyd 98%
Lieferant: Apollo Scientific
Synonyme:
4-Methylthiazole-5-carboxaldehyde, 5-formyl-4-methyl thiazole, 4-Methyl-5-formylthiazole
OR8850-10G
APOSOR8850-10GEA
154
EUR
APOSOR8850-10G
4-Methyl-1,3-thiazol-5-carbaldehyd 98%
4-Methylthiazole-5-carbaldehyde
Formel:
C₅H₅NOS Molecular Weight: 127,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368277 CAS-Nummer: 82294-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...