7-(Benzyloxy)-DL-tryptophan

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-[7-(benzyloxy)-1H-indol-3-yl]propanoic acid

OR8850T-100MG OR8850T-10MG OR8850T-250MG
APOSOR8850T-100MGEA 277 EUR
APOSOR8850T-100MG APOSOR8850T-10MG APOSOR8850T-250MG
7-(Benzyloxy)-DL-tryptophan
7-(Benzyloxy)-DL-tryptophan
Formel: C₁₈H₁₈N₂O₃
Molecular Weight: 310.347 g/mol
Schmelzpunkt: 234.5…236.0 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00037969
CAS-Nummer: 66866-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR