2-Chlor-5-cyanphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR8868-1G
APOSOR8868-1GEA
30.6
EUR
APOSOR8868-1G
2-Chlor-5-cyanphenylboronsäure
2-Chlor-5-cyanphenylboronsäure
Formel:
C₇H₅BClNO₂ Molecular Weight: 181,3841 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06656271 CAS-Nummer: 936249-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...