2-Methoxy-3-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxypyridine-3-boronic acid

OR8870-25G OR8870-5G OR8870-100G
APOSOR8870-25GEA 88.6 EUR
APOSOR8870-25G APOSOR8870-5G APOSOR8870-100G
2-Methoxy-3-pyridinboronsäure
2-Methoxy-3-pyridineboronic acid
Formel: C₆H₈BNO₃
Lagertemperatur: Kühlschrank
MDL: MFCD03411572
CAS-Nummer: 163105-90-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR