2-Amino-3-methoxypyridin 97%

Lieferant: Apollo Scientific
Achtung

OR8879-1G OR8879-5G
APOSOR8879-1GEA 89.9 EUR
APOSOR8879-1G APOSOR8879-5G
2-Amino-3-methoxypyridin 97%
2-Amino-3-methoxypyridine
Formel: C₆H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07374874
CAS-Nummer: 10201-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR