2-(4-Boronophenyl)quinoline-4-carboxylic acid

Lieferant: Apollo Scientific

OR8881-1G
APOSOR8881-1GEA 760 EUR
APOSOR8881-1G
2-(4-Boronophenyl)quinoline-4-carboxylic acid
2-(4-Boronophenyl)quinoline-4-carboxylic acid
Formel: C₁₆H₁₂BNO₄
Lagertemperatur: Kühlschrank
MDL: MFCD01631225
CAS-Nummer: 373384-17-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR