5-Brom-2-methoxybenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(5-Brom-2-methoxyphenyl)methanol, 4-Brom-3-(hydroxymethyl)anisol
OR8893-5G
APOSOR8893-5GEA
66.9
EUR
APOSOR8893-5G
5-Brom-2-methoxybenzylalkohol 98%
5-Brom-2-methoxybenzylalkohol
Formel:
C₈H₉BrO₂ Molecular Weight: 217,06 g/mol Schmelzpunkt: 68…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004630 CAS-Nummer: 80866-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...