2,4,5-Tribromimidazol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,4,5-Tribrom-1H-imidazol
OR8896-100G
APOSOR8896-100GEA
129
EUR
APOSOR8896-100G
2,4,5-Tribromimidazol
2,4,5-Tribromoimidazole
Formel:
C₃HBr₃N₂ Molecular Weight: 304,77 g/mol Schmelzpunkt: 217…220 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00005184 CAS-Nummer: 2034-22-2 EINECS: 217-997-5 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...