5-Isochinolincarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Isoquinoline-5-carboxylic acid
OR8925-1G
OR8925-250MG
APOSOR8925-1GEA
87.3
EUR
APOSOR8925-1G
APOSOR8925-250MG
5-Isochinolincarbonsäure
5-Isoquinolinecarboxylic acid
Formel:
C₁₀H₇NO₂ Molecular Weight: 173.17 g/mol Siedepunkt: 60…62 °C/11mmHg Schmelzpunkt: 275…281 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03788744 CAS-Nummer: 27810-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...