Methyl-5-bromvalerat
Lieferant: Apollo Scientific
OR8928-100G
OR8928-25G
OR8928-500G
APOSOR8928-100GEA
0
EUR
APOSOR8928-100G
APOSOR8928-25G
APOSOR8928-500G
Methyl-5-bromvalerat
Methyl-5-bromvalerat
Formel:
BrH₂C(CH₂)₃COOCH₃ Dichte: 1,363 g/cm³ (25 °C) Flammpunkt: 99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000265 CAS-Nummer: 5454-83-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...