Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-acetamido-2-hydroxybenzoate

OR8935-100G
APOSOR8935-100GEA 60.9 EUR
APOSOR8935-100G
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate 95+%
Methyl-4-(acetylamino)-2-hydroxybenzenecarboxylate
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.2 g/mol
Schmelzpunkt: 146…147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00458326
CAS-Nummer: 4093-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR