4-Brom-3-methylbenzolsulfonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Brom-3-methylbenzolsulfonsäurechlorid, 4-Brom-3-methylbenzol-1-sulfonylchlorid, 4-Brom-3-methylbenzolsulfochlorid
OR8938-25G
APOSOR8938-25GEA
253
EUR
APOSOR8938-25G
4-Brom-3-methylbenzolsulfonylchlorid 97%
4-Brom-3-methylbenzolsulfonylchlorid
Formel:
C₇H₆BrClO₂S Molecular Weight: 269,55 g/mol Schmelzpunkt: 57…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094696 CAS-Nummer: 72256-93-0 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...