4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Chloro-7-chlorosulphonylbenzofurazan, 4-Chloro-2,1,3-benzoxadiazole-7-sulphonyl Chloride
OR8946-1G
APOSOR8946-1GEA
102
EUR
APOSOR8946-1G
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole
4-Chloro-7-chlorosulphonyl-2,1,3-benzoxadiazole
Bifunctional fluorescent reagent. Cross-linking reagent.
Formel:
C₆H₂Cl₂N₂O₃S Molecular Weight: 253,05 g/mol Schmelzpunkt: 86…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191405 CAS-Nummer: 142246-48-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...