6-Chloro-2H-[1,4]benzoxazine-3(4H)-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-2H-benzo[b][1,4]oxazin-3(4H)-one
OR8948-25G
APOSOR8948-25GEA
78.3
EUR
APOSOR8948-25G
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one
6-Chloro-2H-[1,4]benzoxazine-3(4H)-one
Formel:
C₈H₆ClNO₂ Molecular Weight: 183,5917 g/mol Schmelzpunkt: 215…219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00174511 CAS-Nummer: 7652-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...