4-Nitro-1H-pyrazole-3-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitro-3-pyrazolecarboxylic acid, 4-Nitropyrazole-3-carboxylic acid

OR8951-100G
APOSOR8951-100GEA 161 EUR
APOSOR8951-100G
4-Nitro-1H-pyrazole-3-carboxylic acid 98%
4-Nitro-1H-pyrazole-3-carboxylic acid
Formel: C₄H₃N₃O₄
Molecular Weight: 157.085 g/mol
Schmelzpunkt: 223…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00090899
CAS-Nummer: 5334-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR