3'-Phenoxyacetophenon
Lieferant: Apollo Scientific
Synonyme:
1-(3-Phenoxyphenyl)ethan-1-one, 1-(3-Phenoxyphenyl)ethanone
OR8954-1G
APOSOR8954-1GEA
60.9
EUR
APOSOR8954-1G
3'-Phenoxyacetophenon
3'-Phenoxyacetophenon
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,2439 g/mol |
MDL:
MFCD08543460 CAS-Nummer: 32852-92-9 EINECS: 251-259-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...