3-(3-Aminophenyl)propionsäure 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(3-Aminophenyl)propanoic acid

OR8979-1G
APOSOR8979-1GEA 21.9 EUR
APOSOR8979-1G
3-(3-Aminophenyl)propionsäure 95%
3-(3-Aminophenyl)propionsäure
Formel: C₉H₁₁NO₂
Schmelzpunkt: 99…101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00153889
CAS-Nummer: 1664-54-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR