1,2,2,3-Tetrachlorpropan 99%

Lieferant: Apollo Scientific

OR8992-5G
APOSOR8992-5GEA 165 EUR
APOSOR8992-5G
1,2,2,3-Tetrachlorpropan 99%
1,2,2,3-Tetrachloropropane
Formel: ClH₂CCCl₂CH₂Cl
Molecular Weight: 181,88 g/mol
Siedepunkt: 87…88 °C/50mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00039362
CAS-Nummer: 13116-53-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR