2-Chlor-5-hydroxypyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR8994-100G
OR8994-25G
OR8994-500G
APOSOR8994-100GEA
202
EUR
APOSOR8994-100G
APOSOR8994-25G
APOSOR8994-500G
2-Chlor-5-hydroxypyridin 98%
Formel:
C₅H₄ClNO Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234050 CAS-Nummer: 41288-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...