Methyltetrahydropyran-4-ylacetat 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(tetrahydro-2H-pyran-4-yl)acetate

OR900002-5G
APOSOR900002-5GEA 106 EUR
APOSOR900002-5G
Methyltetrahydropyran-4-ylacetat 96%
Methyltetrahydropyran-4-ylacetat
Formel: C₈H₁₄O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09972087
CAS-Nummer: 156002-64-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR