5-Chlor-2-hydroxy-3-nitropyridin 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-3-nitropyridin-2-ol

OR9001-1G
APOSOR9001-1GEA 21.9 EUR
APOSOR9001-1G
5-Chlor-2-hydroxy-3-nitropyridin 95+%
5-Chlor-2-hydroxy-3-nitropyridin
Formel: C₅H₃ClN₂O₃
Molecular Weight: 174,54 g/mol
Schmelzpunkt: 231…234 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00114884
CAS-Nummer: 21427-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR