1-Benzylpyrrolidine-3-carbonitrile 96%
Lieferant: Apollo Scientific
OR900130-5G
APOSOR900130-5GEA
58
EUR
APOSOR900130-5G
1-Benzylpyrrolidine-3-carbonitrile 96%
1-Benzylpyrrolidine-3-carbonitrile
Formel:
C₁₂H₁₄N₂ Molecular Weight: 186.26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05662780 CAS-Nummer: 10603-52-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...