3-(Trimethylsilyl)-2-propin-1-ol 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Trimethylsilyl)propargyl alcohol

OR900149-100G
APOSOR900149-100GEA 169 EUR
APOSOR900149-100G
3-(Trimethylsilyl)-2-propin-1-ol 98%
3-(Trimethylsilyl)-2-propin-1-ol
Formel: C₆H₁₂OSi
Molecular Weight: 128.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00002913
CAS-Nummer: 5272-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR