3-(2-Chlorophenyl)propan-1-ol 98%

Lieferant: Apollo Scientific

OR900157-1G
APOSOR900157-1GEA 270 EUR
APOSOR900157-1G
3-(2-Chlorophenyl)propan-1-ol 98%
3-(2-Chlorophenyl)propan-1-ol
Formel: C₉H₁₁ClO
Molecular Weight: 170,636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00093567
CAS-Nummer: 6282-87-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR