2-Cyclohepten-1-one 90%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Cyclohept-2-enone
OR900263-25G
APOSOR900263-25GEA
145
EUR
APOSOR900263-25G
2-Cyclohepten-1-one 90%
2-Cyclohepten-1-one
Formel:
C₇H₁₀O Molecular Weight: 110.16 g/mol Dichte: 0.988 g/cm³ (25 °C) Flammpunkt: 68 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004157 CAS-Nummer: 1121-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...