4-Ethoxybenzonitril 98%

Lieferant: Apollo Scientific

Synonyme: p-Ethoxybenzonitrile

OR900340-100G
APOSOR900340-100GEA 186 EUR
APOSOR900340-100G
4-Ethoxybenzonitril 98%
4-Ethoxybenzonitril
Formel: C₉H₉NO
Molecular Weight: 147.18 g/mol
Siedepunkt: 258
Lagertemperatur: Raumtemperatur
MDL: MFCD00001819
CAS-Nummer: 25117-74-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR