4-(2-Methylpropoxy)benzonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
4-iso-Butoxybenzonitrile
OR900341-1G
OR900341-5G
APOSOR900341-1GEA
40.6
EUR
APOSOR900341-1G
APOSOR900341-5G
4-(2-Methylpropoxy)benzonitrile 98%
4-(2-Methylpropoxy)benzonitrile
Formel:
C₁₁H₁₃NO Molecular Weight: 175.227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08274895 CAS-Nummer: 5203-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...